Please login to your online account to display your discounted pricing

RJR 2403 oxalate, Tocris


α4β2 selective nicotinic agonist

Quantity 10mg

Manufacturer: r & d systems  RDS105310

 View more versions of this product

Catalog No. 105310

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 252.27
Quantity 10mg
Molecular Formula C10H14N2·C2H2O4
Chemical Name or Material RJR 2403 oxalate
Synonym (E)-N-Methyl-4-(3-pyridinyl)-3-buten-1-amine oxalate
CAS 220662-95-3
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material RJR 2403 oxalate
Assay Percent Range >99%
Molecular Formula C10H14N2·C2H2O4
Formula Weight 252.27
Synonym (E)-N-Methyl-4-(3-pyridinyl)-3-buten-1-amine oxalate
Accessories

Desiccate at +4°C