Please login to your online account to display your discounted pricing

RJR 2403 oxalate, Tocris

α4β2 selective nicotinic agonist

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material RJR 2403 oxalate
Assay Percent Range >99%
Molecular Formula C10H14N2·C2H2O4
Formula Weight 252.27
Synonym (E)-N-Methyl-4-(3-pyridinyl)-3-buten-1-amine oxalate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

105350

 
r & d systems
RDS105350
50mg Each for $682.67

105310

 
r & d systems
RDS105310
10mg Each for $164.61
Description & Specifications

Specifications

Formula Weight 252.27
Quantity 50mg
Chemical Name or Material RJR 2403 oxalate
CAS 220662-95-3
Molecular Formula C10H14N2·C2H2O4
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C
Synonym (E)-N-Methyl-4-(3-pyridinyl)-3-buten-1-amine oxalate
Safety and Handling

Desiccate at +4°C