missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rizatriptan Benzoate 98.0+%, TCI America™

Manufacturer:  Tci America R018150MG

 View more versions of this product

Catalog No. R018150MG



Specifications

Specifications

Rizatriptan Benzoate
180°C
145202-66-0
MFCD00866224
JPRXYLQNJJVCMZ-UHFFFAOYSA-N
benzoic acid;N,N-dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine
77997
Crystal-Powder at 20°C
White
50mg
C22H25N5O2
2-5-1h-1,2,4-triazol-1-yl methyl-1h-indol-3-yl-n,n-dimethylethanamine benzoate, maxalt, maxalt-mlt, n,n-dimethyl-2-5-1,2,4-triazol-1-ylmethyl-1h-indol-3-yl ethylamine benzoate, rizatrimptan benzoate, rizatriptan benzoate, rizatriptan benzoate usan:usp, rizatriptane benzoate, unii-wr978s7qhh
CN(C)CCC1=CNC2=C1C=C(C=C2)CN3C=NC=N3.C1=CC=C(C=C1)C(=O)O
391.475
391.48
≥98.0% (HPLC,T)

Chemical Identifiers

145202-66-0
391.475
JPRXYLQNJJVCMZ-UHFFFAOYSA-N
77997
CN(C)CCC1=CNC2=C1C=C(C=C2)CN3C=NC=N3.C1=CC=C(C=C1)C(=O)O
C22H25N5O2
MFCD00866224
2-5-1h-1,2,4-triazol-1-yl methyl-1h-indol-3-yl-n,n-dimethylethanamine benzoate, maxalt, maxalt-mlt, n,n-dimethyl-2-5-1,2,4-triazol-1-ylmethyl-1h-indol-3-yl ethylamine benzoate, rizatrimptan benzoate, rizatriptan benzoate, rizatriptan benzoate usan:usp, rizatriptane benzoate, unii-wr978s7qhh
benzoic acid;N,N-dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethanamine
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions