missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Risperidone 98.0+%, TCI America™

Manufacturer:  TCI America R0087500MG

 View more versions of this product

Catalog No. R0087500MG



Specifications

Specifications

Risperidone
172°C
106266-06-2
MFCD00274576
risperidone, risperidal, risperdal, risperdal consta, rispolept, risperin, rispolin, sequinan, risperidona, risperidonum
CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
410.493
CHEBI:8871
Crystal-Powder at 20°C
White-Yellow
500mg
C23H27FN4O2
2811
RAPZEAPATHNIPO-UHFFFAOYSA-N
3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
5073
410.49
≥98.0% (HPLC,T)

Chemical Identifiers

106266-06-2
410.493
RAPZEAPATHNIPO-UHFFFAOYSA-N
5073
3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
C23H27FN4O2
MFCD00274576
risperidone, risperidal, risperdal, risperdal consta, rispolept, risperin, rispolin, sequinan, risperidona, risperidonum
CHEBI:8871
CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
Safety and Handling

Safety and Handling

RTECSNumber : UV1164800

TSCA : No

Documents