Ripple Tank High-Power Light Source
Makes ripple tank operation more convenient and produces sharper images.
Manufacturer:  Fisher Scientific BMCO14700
 View more versions of this product
Catalog No. S52062
Specifications Description & Specifications
Specifications
High power light source
150w
Power cord, socket, bulb, shield, spring clamp