Please login to your online account to display your discounted pricing
Science First™ Ripple Tank High-Power Light Source
Makes ripple tank operation more convenient and produces sharper images.
$46.95 - $129.90
Specifications
Includes Power cord, socket, bulb, shield, spring clamp
Wattage 150w
View More Specs
Includes:  Power cord, socket, bulb, shield, spring clamp
Products
Catalog Number Mfr. No. Product Type Price Quantity    
S52062A
science first™
1470001P
Replacement bulb Each for $46.95
S52062
fisher scientific
BMCO14700
High power light source Each for $129.90
Description & Specifications
Specifications
Includes Power cord, socket, bulb, shield, spring clamp
Wattage 150w
  • 150w clear bulb mounted inside a fully vented flat black shield
  • Lamp shield rotates to provide a point source or a straight line source