missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human RIPK2, His Tag Recombinant Protein
RIPK2 is a serine/threonine kinase.
$768.00 - $11260.00
Specifications
Accession Number | NP_003812 |
---|---|
Molecular Weight (g/mol) | 38.9 kDa |
Gene Alias | RIPK2, RICK, RIP2 |
KinaseFamily | RIPK Kinase Family |
Species | Human |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR7829A
|
Invitrogen™
PR7829A |
100μg |
Each for $2,645.00
|
|
|||||
PV4325
|
Invitrogen™
PV4325 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PV4213
|
Invitrogen™
PV4213 |
10 μg |
Each for $768.00
|
|
|||||
Description
RIPK2 is involved in cell activation, proliferation, differentiation, and apoptosis.
Specifications
NP_003812 | |
RIPK2, RICK, RIP2 | |
Human | |
His Tag | |
Ser⁄Thr Kinases: TKL Group | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
38.9 kDa | |
RIPK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |