Invitrogen™ RIPK2 Recombinant Human Protein DFS Item
RIPK2 is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4325
 View more versions of this product
Catalog No. PV4325
Description
Description
RIPK2 is involved in cell activation, proliferation, differentiation, and apoptosis.
Specifications
Specifications
NP_003812
Frozen
38.9kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
RIPK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
RIPK2, RICK, RIP2
Baculovirus
Human
His Tag
Documents