missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ RIPA buffer

$90.22 - $90.22

Specifications

Chemical Name or Material RIPA buffer
MDL Number MFCD02100484
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63306AK View Documents Alfa Aesar™
J63306AK
250mL Each for $90.22
 
AAJ63306AP View Documents Alfa Aesar™
J63306AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
RIPA is used as a high quality, ready to use mammalian cell and tissue lysis buffer with protease inhibitors included, is used to lyse cells and tissue, for radio immunoprecipitation assay.

Solubility
Fully miscible.

Notes
50mM Tris-HCl (pH 7.4), 150mM NaCl 1% NP-40, 0.5% sodium deoxycholate and 0.1% SDS
Specifications

Specifications

RIPA buffer
MFCD02100484