Please login to your online account to display your discounted pricing

Rink amide benzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5mmol/g, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  349290010

 View more versions of this product

Catalog No. AC349290010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Off-white to pinkish-brown
Physical Form Beads
Quantity 1g
Additional Information crosslinking: 1% DVB
Chemical Name or Material Rink amide benzhydrylamine polymer resin
MDL Number MFCD00677979
Loading 0.5 to 0.9mmol/g
Mesh Size 200 to 400 mesh
Packaging Glass bottle

Specifications

Chemical Name or Material Rink amide benzhydrylamine polymer resin
MDL Number MFCD00677979