Please login to your online account to display your discounted pricing

Rink amide benzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5mmol/g, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Rink amide benzhydrylamine polymer resin
MDL Number MFCD00677979
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC349290050

  SDS
thermo fisher scientific
349290050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC349290010

  SDS
thermo fisher scientific
349290010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Off-white to pinkish-brown
Physical Form Beads
Quantity 5g
Packaging Glass bottle
Additional Information crosslinking: 1% DVB
Chemical Name or Material Rink amide benzhydrylamine polymer resin
MDL Number MFCD00677979
Loading 0.5 to 0.9mmol/g
Mesh Size 200 to 400 mesh

for solid support synthesis