Please login to your online account to display your discounted pricing

Rink amide,4-methylbenzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5mmol/g, Acros Organics


Quantity 5g

Manufacturer: thermo fisher scientific  349310050

 View more versions of this product

Catalog No. AC349310050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Beads
Color Off-white to pinkish-brown
Quantity 5g
MDL Number MFCD00677978
Loading 0.5 to 0.9mmol/g
Additional Information crosslinking: 1% DVB
Mesh Size 200 to 400 mesh
Chemical Name or Material Rink amide, 4-methylbenzhydrylamine polymer resin

Specifications

Chemical Name or Material Rink amide, 4-methylbenzhydrylamine polymer resin
MDL Number MFCD00677978