Please login to your online account to display your discounted pricing

Rink amide,4-methylbenzhydrylamine polymer resin, 1% crosslinked with DVB, 0.5mmol/g, Acros Organics

Chemical Identifiers

Chemical Name or Material Rink amide, 4-methylbenzhydrylamine polymer resin
MDL Number MFCD00677978
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC349310010

  SDS
thermo fisher scientific
349310010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC349310050

  SDS
thermo fisher scientific
349310050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Off-white to pinkish-brown
Physical Form Beads
Quantity 1g
Additional Information crosslinking: 1% DVB
MDL Number MFCD00677978
Loading 0.5 to 0.9mmol/g
Mesh Size 200 to 400 mesh
Chemical Name or Material Rink amide, 4-methylbenzhydrylamine polymer resin

for solid support synthesis