Please login to your online account to display your discounted pricing

Right to Know SDS Center


Centralized access to SDS as outlined in OSHA Right-to-Know standards.

Manufacturer: fisher science education  2023EMD

 View more versions of this product

Catalog No. S47471

  • / Each
 

Description & Specifications

Specifications

Format Binder holds 275 pages
Category Safety and Training Materials
For Use With (Application) Provides workers with easy access to material safety data sheets with the right-to-know compliance center
Product Type MSDS Binder; H x W x D: 11.5 x 10.5 x 1.5 in.
Includes 1.5 in. diameter rings and 36 in. chain