Please login to your online account to display your discounted pricing

Right to Know SDS Center


Centralized access to SDS as outlined in OSHA Right-to-Know standards.

Manufacturer: fisher science education  2010EMD

 View more versions of this product

Catalog No. S47470

  • / Each
 

Description & Specifications

Specifications

Format Center with binder holds 275 pages
Category Safety and Training Materials
For Use With (Application) Provides workers with easy access to material safety data sheets with the right-to-know compliance center
Product Type Wall Mount Right-To-Know Compliance Center; H x W x D: 20 x 14 x 4.5 in. with Binder.
Includes Board, Wire Binder Holder, and MSDS Binder