Please login to your online account to display your discounted pricing

Rifampicin, Fisher BioReagents


Quantity 25g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP267925

 View more versions of this product

Catalog No. BP267925

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 822.96
Melting Point 183°C
Quantity 25g
Merck Index 15, 8341
Synonym Rif
Physical Form Solid
Molecular Formula C43H55N4O12
Color Orange
Chemical Name or Material Rifampicin
Identification Pass Test
Heavy Metals < 20ppm
Assay Percent Range 97%
ChemAlert Storage Symbol Gray
One Peak < 1.0%
Health Hazard 3 Emergency Overview
Substances known to cause developmental toxicity in humans. Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:2
Flammability:0
Instability:0
Recommended Storage 4°C
Loss on Drying < 1.0%
Packaging Amber Glass
Purity Inclusive between 97.0%
Rifampicin quinine < 1.5%
Health Hazard 2 WARNING!
Sulfated Ash < 0.1%
Sum of Peaks < 3.5%
Crystallinity Pass Test
CAS 13292-46-1

Specifications

Chemical Name or Material Rifampicin
CAS 13292-46-1
Physical Form Solid
Assay Percent Range 97%
Molecular Formula C43H55N4O12
Formula Weight 822.96
Synonym Rif

  • An antibiotic used to inhibit synthesis of proteins during recombinant protein expression in bacteria
  • Rif inhibits bacterial DNA-dependent RNA polymerase
  • Rif can also be used as a selective agent for some strains
  • Mammalian RNA polymerase is not affected

Accessories
 

Emergency Overview
Substances known to cause developmental toxicity in humans. Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

4°C

SDS & Safety Documents