Please login to your online account to display your discounted pricing
Thermo Scientific™ Richard-Allan Scientific™ Steiner-Steiner Silver
Determine the presence of spirochetes and nonfilamentous bacteria with Thermo Scientific™ Richard-Allan Scientific™ Steiner-Steiner Silver.
$759.47 - $759.47
Products
Catalog Number Mfr. No. Type Price Quantity    
23-900-657
thermo scientific™
87009
Steiner-Steiner Silver Kit Each for $759.47
Description & Specifications
Determine the presence of spirochetes and nonfilamentous bacteria with Thermo Scientific™ Richard-Allan Scientific™ Steiner-Steiner Silver.
  • Kit does not contain uranyl nitrate
  • Background is stained yellow to brown
  • Bacteria, Donovan bodies and fungi all stain black
  • Kit formulation and detailed procedures provide consistent and reproducible results