missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Signature Series™ Hematoxylin 7211
Yield results that exceed those of Gill or Harris Hematoxylins with this uniquely formulated product.
$573.26 - $2482.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
22-050-111
|
Epredia™
7211 |
1 pt. |
Case of 4 Each for $573.26
|
|
|||||
72-11L
|
Epredia™
7211L |
1.32 gal. |
Each for $2,482.00
|
|
|||||
Description
- No filtration required
- Stains up to 2500 slides per pint
- Crisp, well-defined nuclear detail
Available in 5 liter packaging
- Reduces plastic by 90%
- Increases stain stability
- Reduces messy stain spills
- Saves 35% storage space versus traditional packaging
Specifications
Hematoxylin |