Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives
Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
Manufacturer:  Thermo Scientific™ 152460M
 View more versions of this product
Catalog No. 152460M
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Use Thermo Scientific™ Richard-Allan Scientific™ Reusable Steel Knives with microtomes or cryostats when the technical application dictates that requirement.
  • Permanent knives can be resharpened on an appropriate knife sharpener for extended use
  • Appropriate knife case(s) should be ordered with the knife or knives, each case holds two knives
Multiple Styles Available:
  • C-profile knives are used for standard paraffin and cryo techniques
  • D-profile knives are used for sectioning of harder materials that require greater knife stability
  • Tungsten carbide knives are used for strength and durability in sectioning of very hard samples or embedded resin
Specifications
Specifications
Richard-Allan Scientific™
Steel Knives
16cm
Solid Tungsten
D
No
D-Profile Solid Tungsten; 16cm
Documents