Please login to your online account to display your discounted pricing
Thermo Scientific™ Richard-Allan Scientific™ Mucicarmine
Identify primary tumor sites with the aid of Thermo Scientific™ Richard-Allan Scientific™ Mucicarmine
$762.19 - $762.19
Products
Catalog Number Mfr. No. Type Price Quantity    
22-110-646
thermo scientific™
87012
Mucicarmine stain kit Each for $762.19
Description & Specifications
Identify primary tumor sites with the aid of Thermo Scientific™ Richard-Allan Scientific™ Mucicarmine
  • Aids in distinguishing mucin-negative undifferentiated squamous cell lesions from mucin-positive adenocarcinomas
  • Mucin is stained dark rose and the nuclei are black
  • Capsule of Cryptococcus (fungus) is stained deep rose
  • Tartrazine solution is used as a yellow cytoplasmic stain