Please login to your online account to display your discounted pricing
Thermo Scientific™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen)
Save time with Thermo Scientific™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome.
$702.56 - $702.56
Products
Catalog Number Mfr. No. Type Price Quantity    
23-900-662
thermo scientific™
87020
Stain Kit Each for $702.56
Description & Specifications
Save time with Thermo Scientific™ Richard-Allan Scientific™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome.
  • Used to study the diseases of connective tissue and muscle associated with fibrotic and dystrophic changes
  • Stains collagen blue, cytoplasm and muscle fibers red and the nuclei blue-to-black