Please login to your online account to display your discounted pricing
Thermo Scientific Richard-Allan Scientific™ Fine Cutting Tool Kit
Improve consistency in grossing with the Thermo Scientific™ Richard-Allan Scientific™ Fine Cutting Tool Kit.
$1,518.44 - $1,525.79
Products
Catalog Number Mfr. No. Description Price Quantity    
98-13
thermo fisher scientific
9813
Fine Cutting Tool Kit, 3mm Each for $1,518.44
98-15
thermo fisher scientific
9815
Fine Cutting Tool Kit, 5mm Each for $1,525.79
Description & Specifications
Improve consistency in grossing with the Thermo Scientific™ Richard-Allan Scientific™ Fine Cutting Tool Kit.
Features:
  • Easy to clean
  • Chemical resistant
  • Autoclavable
Ordering information:
  • Choose the correct tool to gross specimens at 3, 4 or 5mm thickness.
  • Includes hex wrench and a 10-pack of high profile disposable blades.