Please login to your online account to display your discounted pricing
Thermo Scientific™ Richard-Allan Scientific™ Amyloid Stain
Demonstrate amyloid with Thermo Scientific™ Richard-Allan Scientific™ Amyloid Stain.
$494.50 - $494.50
Products
Catalog Number Mfr. No. Type Price Quantity    
23-900-655
thermo scientific™
87003
Stain Kit Each for $494.50
Description & Specifications
Demonstrate amyloid with Thermo Scientific™ Richard-Allan Scientific™ Amyloid Stain.
  • Alkaline Congo Red solution will stain amyloid orange-to-red and display apple-green birefringence when examined with polarizing microscope