Invitrogen™ RiboMinus™ Pan-Prokaryote Probe Mix DFS Item
The RiboMinus™ Pan-Prokaryote Probe Mix is a pool of more than 150 3′-biotin labeled ssDNA oligonucleotides that are used to capture and eliminate bacterial and archaebacterial ribosomal RNA from RNA samples in preparation for NGS transcriptome studies.
Supplier:  Invitrogen™ A46920
Catalog No. A46920
Description
Description
RiboMinus™ Pan-Prokaryote Probe Mix is a component of the RiboMinus Bacteria 2.0 Transcriptome Isolation Kit and is available as a standalone product for use in conjunction with other RiboMinus products targeting other species: Plant (Cat. No. A1083808) and Eukaryote v2 (Cat. No. A15026). Combining RiboMinus Pan-Prokaryote Probe Mix with Plant or Eukaryote probe mixes is useful for metatranscriptomic studies. This probe mix consists of over 150 3’-biotin-labeled ssDNA oligonucleotide designs. These designs were created using highly conserved regions of rRNA across 76 pan-prokaryote species (see table below) and tested with several species (see compatibility list below).
Specifications
Specifications
RiboMinus Isolation kits include RiboMinus Magnetic Beads, RiboMinus human/mouse, bacteria, or yeast probe, and Hybridization Buffer. The optional RiboMinus Concentration Module contains spin cartridges, wash tubes, recovery tubes, Binding Buffer, Wash Buffer, and RNase-free (DEPC-treated) water. Store the isolation modules at +4°C and the concentration module at room temperature. All reagents are guaranteed stable for 6 months when properly stored.
Magnetic Bead
Total RNA (Bacterial)
Not High-throughput Compatible (Manual)
12
1
Transcriptome RNA
Magnetic Bead
1.25 hr.
Transcriptome RNA
Next Generation Sequencing, Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction
RiboMinus™
100 ng to 5 ug of total RNA
RiboMinus
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.