Please login to your online account to display your discounted pricing

Alfa Aesar™ Riboflavin, 98%, Electrophoresis Grade

$68.26 - $68.26

Chemical Identifiers

Chemical Name or Material Riboflavin
CAS 83-88-5
Purity Grade Electrophoresis Grade
Formula Weight 376.37
MDL Number MFCD00005022
Merck Index 14,8200
Synonym Vitamin B2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6663214

  SDS
alfa aesar™
J6663214
25g Each for $68.26
Description & Specifications

Specifications

Formula Weight 376.37
Chemical Name or Material Riboflavin
CAS 83-88-5
TSCA TSCA
MDL Number MFCD00005022
Melting Point 290°C
Physical Form Powder
Purity Grade Electrophoresis Grade
Merck Index 14,8200
Synonym Vitamin B2
RTECS Number VJ1400000
Color Orange-yellow
Sensitivity Light Sensitive
Storage Note 1 2° to 8°C
Quantity 25g
Odor Odorless
Assay 98%
Safety and Handling

2° to 8°C