missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Riboflavin 5′-Monophosphate Sodium Salt, TCI America™

Manufacturer:  TCI America R002325G

 View more versions of this product

Catalog No. R002325G



Specifications

Specifications

Riboflavin 5′-Monophosphate Sodium Salt
25g
C17H23N4NaO9P
Flavin Mononucleotide, Lactoflavin Phosphate Sodium Salt, Vitamin B2 Phosphate Sodium Salt
[HH].CC1=CC2=C(C=C1C)N(C3=NC(=O)NC(=O)C3=N2)CC(C(C(COP(=O)(O)O)O)O)O.[Na]
481.354
478.33
≥93.0% (E)
Undesignated
130-40-5
MFCD00065362
IJRXPIRQLSSYGH-APQIITSESA-N
[(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;molecular hydrogen;sodium
131673928
Crystal-Powder at 20°C

Chemical Identifiers

130-40-5
481.354
IJRXPIRQLSSYGH-APQIITSESA-N
131673928
[HH].CC1=CC2=C(C=C1C)N(C3=NC(=O)NC(=O)C3=N2)CC(C(C(COP(=O)(O)O)O)O)O.[Na]
C17H23N4NaO9P
MFCD00065362
Flavin Mononucleotide, Lactoflavin Phosphate Sodium Salt, Vitamin B2 Phosphate Sodium Salt
[(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;molecular hydrogen;sodium
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

Documents