Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Rhodopsin (Opsin) Ab-1, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Rhodopsin (Opsin) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1233P1ABX

 View more versions of this product

Catalog No. MS1233P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Membrane preparation from adult rat retina
Regulatory Status RUO
Clone RET-P1
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Mouse
Isotype IgG1
Antigen Rhodopsin (Opsin) Ab-1
Format Concentrated
Quantity 100µL
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cancer and Tumor Biology
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Classification Monoclonal
Item Description 100uL; 1mg/mL; Unlabeled; Purified without BSA and azide

Provide accurate, reproducible results with the Thermo Scientific™ Rhodopsin (Opsin) Ab-1, Mouse Monoclonal Antibody.

Rhodopsin is a glycoprotein which is a visual pigment in mammals, birds, amphibians, and reptiles. The rhodopsin protein is present on the photoreceptor cells in the retina.

Host Species: Mouse

Clone: RET-P1

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Membrane preparation from adult rat retina

Molecular Weight: 39kDa

Positive Control: Brain or retina

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO