Please login to your online account to display your discounted pricing

Rhodizonic acid, disodium salt dihydrate, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC382710050

 View more versions of this product

Catalog No. AC382710050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 250.07
Quantity 5g
CAS 166316-02-5
Chemical Name or Material Rhodizonic acid, disodium salt dihydrate
Packaging Glass bottle
MDL Number MFCD06658965
Molecular Formula C6O6Na2·2H2O

Specifications

Chemical Name or Material Rhodizonic acid, disodium salt dihydrate
CAS 166316-02-5
Molecular Formula C6O6Na2·2H2O
Formula Weight 250.07
MDL Number MFCD06658965