missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodizonic acid, disodium salt dihydrate, ACROS Organics™

Chemical Identifiers

CAS 166316-02-5
Molecular Formula C6O6Na2·2H2O
Formula Weight 250.07
MDL Number MFCD06658965
Chemical Name or Material Rhodizonic acid, disodium salt dihydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC382710050 SDS Thermo Fisher Scientific
AC382710050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC382710010 SDS Thermo Fisher Scientific
AC382710010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

166316-02-5
250.07
Rhodizonic acid, disodium salt dihydrate
C6O6Na2·2H2O
MFCD06658965

Specifications

250.07
166316-02-5
MFCD06658965
Rhodizonic acid, disodium salt dihydrate
5g
Glass bottle
C6O6Na2·2H2O