Please login to your online account to display your discounted pricing

Rhodizonic acid, disodium salt, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  132340050

 View more versions of this product

Catalog No. AC132340050

  • / Each
 

Description & Specifications

Chemical Identifiers

Appearance of Solution (0.2% aq. soln.) clear
Chemical Name or Material Rhodizonic acid, disodium salt
Additional Information Suitable for determination of barium
CAS 523-21-7
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Fine Crystalline Powder
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6O6Na2
Formula Weight 214.04
Synonym Sodium rhodizonate
Color Green-gray to dark green
Quantity 5g

Specifications

Chemical Name or Material Rhodizonic acid, disodium salt
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6O6Na2
Formula Weight 214.04
Synonym Sodium rhodizonate