Please login to your online account to display your discounted pricing

Rhodizonic acid, disodium salt, 98%, ACROS Organics™

$44.74 - $585.68

Chemical Identifiers

Chemical Name or Material Rhodizonic acid, disodium salt
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6O6Na2
Formula Weight 214.04
Synonym Sodium rhodizonate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC132340050

  SDS
fisher scientific acros
132340050
Glass bottle 5g Each for $44.74

AC132340250

  SDS
thermo fisher scientific
AC132340250
Glass bottle 25g Each for $186.13

AC132341000

  SDS
thermo fisher scientific
AC132341000
Glass bottle 100g Each for $585.68
Description & Specifications

Specifications

Appearance of Solution (0.2% aq. soln.) clear
Chemical Name or Material Rhodizonic acid, disodium salt
Additional Information Suitable for determination of barium
CAS 523-21-7
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Fine Crystalline Powder
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6O6Na2
Formula Weight 214.04
Synonym Sodium rhodizonate
Color Green-gray to dark green
Quantity 5g

Indicator