missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rhodizonic acid, disodium salt, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC132340250

 View more versions of this product

Catalog No. AC132340250


 

Specifications Description & Specifications

Specifications

(0.2% aq. soln.) clear
Suitable for determination of barium
Authentic
Fine Crystalline Powder
98%
214.04
Green-gray to dark green
Rhodizonic acid, disodium salt
523-21-7
Glass bottle
≥97.5%
C6O6Na2
Sodium rhodizonate
25g

Chemical Identifiers

98%
214.04
Rhodizonic acid, disodium salt
C6O6Na2
Sodium rhodizonate
≥97.5%