Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodizonic acid disodium salt, 98%

$241.08 - $950.77

Chemical Identifiers

Chemical Name or Material Rhodizonic acid disodium salt
CAS 523-21-7
Molecular Formula C6Na2O6
Formula Weight 214.04
MDL Number MFCD00001650
Beilstein 5181379
Merck Index 14,8670
Synonym 1,2-Dihydroxy-3,4,5,6-tetraoxocyclohexene disodium salt; Sodium rhodizonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1612614

  SDS
alfa aesar™
A1612614
25g Each for $241.08

AAA1612622

  SDS
alfa aesar™
A1612622
100g Each for $950.77

AAA1612606

  SDS
alfa aesar™
A1612606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Rhodizonic acid disodium salt
CAS 523-21-7
Assay 98%
Molecular Formula C6Na2O6
Formula Weight 214.04
MDL Number MFCD00001650
Beilstein 5181379
Merck Index 14,8670
Synonym 1,2-Dihydroxy-3,4,5,6-tetraoxocyclohexene disodium salt; Sodium rhodizonate
Melting Point >300°C
Quantity 25g
Odor Odorless

Applications
Determining sulfate radical by barium chloride titration with rhodizonic acid disodium salt test paper as indicator has been established. Staining of thin sections for quick identification of quartz, potash feldspar and plagioclase is usually done with sodium hexanitrocobaltate (III) and rhodizonic acid disodium salt. N-allyl-N′-(sodium p-phenyl-sulfonate)thiourea is used for eliminating the interference of Ag +?Au 3+?Hg 2+in the identification of Sr 2+ using Rhodizonic acid disodium salt.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.