Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodizonic acid dihydrate, 98%

$64.80 - $64.80

Chemical Identifiers

Chemical Name or Material Rhodizonic acid dihydrate
CAS 63183-44-8
Molecular Formula C6H2O6·2H2O
Formula Weight 206.11 (170.09 Anhydrous)
MDL Number MFCD00149072
Beilstein 2047007
Synonym 2,3,5,5,6,6-Hexahydroxy-2-cyclohexene-1,4-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1246003

 
alfa aesar™
L1246003
1g Each for $64.80
Description & Specifications

Specifications

Formula Weight 206.11 (170.09 Anhydrous)
Chemical Name or Material Rhodizonic acid dihydrate
Assay 98%
Molecular Formula C6H2O6·2H2O
MDL Number MFCD00149072
CAS 63183-44-8
Synonym 2,3,5,5,6,6-Hexahydroxy-2-cyclohexene-1,4-dione
Quantity 1g
Beilstein 2047007
Melting Point ≈248°C

Solubility
Soluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.