missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium wire, 0.5mm (0.02 in.) dia., 99.8% (metals basis)

$181.61 - $3,614.59

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.906
MDL Number MFCD00011201
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11576BR
View Documents
Alfa Aesar™
11576BR
7.5cm Each for $181.61
 
AA11576BS
View Documents
Alfa Aesar™
11576BS
10cm Each for $205.72
 
AA11576BV
View Documents
Alfa Aesar™
11576BV
30cm Each for $541.62
 
AA11576BW
View Documents
Alfa Aesar™
11576BW
50cm Each for $801.99
 
AA11576CB
View Documents
Alfa Aesar™
11576CB
250cm Each for $3,614.59
 
Specifications

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
CHEBI:33359
[Rh]

Specifications

≈2.44g/m
7440-16-6
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Wire, ≈2.44g/m
Rhodium wire, 0.5mm (0.02 in.) dia.
7.5cm
Rh
black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
[Rh]
102.906
CHEBI:33359
99.8%
(metals basis)
Safety and Handling

Safety and Handling

EINECSNumber : 231-125-0

TSCA : Yes

Recommended Storage : Ambient temperatures