missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium wire, 0.5mm (0.02 in.) dia., 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 11576BV

 View more versions of this product

Catalog No. AA11576BV



Specifications

Specifications

Rhodium wire
30cm
7440-16-6
Rh
black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
[Rh]
102.906
CHEBI:33359
99.8%
≈2.44g/m
0.5mm (0.02 in.) dia.
(metals basis)
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Wire, ≈2.44g/m

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
black, dmk383dsac, fume, on alumina, on carbon, rh/al2o3, rhodium, elemental, rhodium, metal, rodio, unii-dmk383dsac
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

EINECSNumber : 231-125-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents