Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium wire, 0.127mm (0.005 in.) dia., 99.8% (metals basis)

Chemical Identifiers

Chemical Name or Material Rhodium wire
CAS 7440-16-6
Name Note 0.127mm (0.005 in.) dia.
MDL Number MFCD00011201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA11578BY

 
alfa aesar™
11578BY
100cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Rhodium wire
TSCA TSCA
Name Note 0.127mm (0.005 in.) dia.
Physical Form Wire
Assay 99.8%
Assay Percent Notes (metals basis)
MDL Number MFCD00011201
CAS 7440-16-6
Weight ≈0.16g/m
Quantity 100cm