missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium wire, 0.127mm (0.005 in.) dia., 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 11578BY

 View more versions of this product

Catalog No. AA11578BY



Specifications

Specifications

Rhodium wire
100cm
7440-16-6
Rh
black
[Rh]
102.906
CHEBI:33359
99.8%
≈0.16g/m
0.127mm (0.005 in.) dia.
(metals basis)
MFCD00011201
MHOVAHRLVXNVSD-UHFFFAOYSA-N
rhodium
23948
Wire

Chemical Identifiers

7440-16-6
102.906
MHOVAHRLVXNVSD-UHFFFAOYSA-N
23948
rhodium
Rh
MFCD00011201
black
CHEBI:33359
[Rh]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents