Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium powder, -22 mesh, Premion™, 99.97% (metals basis), precious metals typically <20ppm each

Chemical Identifiers

Chemical Name or Material Rhodium powder
CAS 7440-16-6
Name Note Precious metals typically <20ppm each
Purity Grade Premion™
MDL Number MFCD00011201
UN Number UN3089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1066801

 
alfa aesar™
1066801
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1066803

 
alfa aesar™
1066803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Rhodium powder
TSCA TSCA
Mesh Size −22 Mesh
Odor Odorless
Name Note Precious metals typically <20ppm each
Purity Grade Premion™
Physical Form Powder
Assay 99.97%
Assay Percent Notes (metals basis)
MDL Number MFCD00011201
CAS 7440-16-6
UN Number UN3089
Color Gray
Quantity 0.25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
RTECS Number VI9069000
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.