missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(IV) oxide, Rh 75.2-77.4%

Manufacturer:  Alfa Aesar™ 4700203

 View more versions of this product

Catalog No. AA4700203



Specifications

Specifications

Rhodium(IV) oxide
O2Rh
rhodium iv oxide, rhodium oxide rho2, rhodium dioxide, rhodium iv oxide, rho2
O=[Rh]=O
134.904
134.9
Rh 75.2 to 77.4%
Hygroscopic
1g
12137-27-8
MFCD00016312
KZYDBKYFEURFNC-UHFFFAOYSA-N
dioxorhodium
82936
Powder
Odorless
Black

Chemical Identifiers

12137-27-8
134.904
KZYDBKYFEURFNC-UHFFFAOYSA-N
82936
O=[Rh]=O
O2Rh
MFCD00016312
rhodium iv oxide, rhodium oxide rho2, rhodium dioxide, rhodium iv oxide, rho2
dioxorhodium
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents