Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(IV) oxide, Rh 75.2-77.4%

Chemical Identifiers

Chemical Name or Material Rhodium(IV) oxide
CAS 12137-27-8
Formula Weight 134.9
MDL Number MFCD00016312
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4700203

  SDS
alfa aesar™
4700203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA47002MD

  SDS
alfa aesar™
47002MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.9
Chemical Name or Material Rhodium(IV) oxide
TSCA TSCA
Odor Odorless
Physical Form Powder
Sensitivity Hygroscopic
MDL Number MFCD00016312
CAS 12137-27-8
Color Black
Quantity 1g
Concentration Rh 75.2 to 77.4%