Alfa Aesar™ Rhodium(IV) oxide, Rh 75.2-77.4%
Manufacturer:  Alfa Aesar™ 47002MD
 View more versions of this product
Catalog No. AA47002MD
Specifications
Specifications
Rhodium(IV) oxide
O2Rh
rhodium iv oxide, rhodium oxide rho2, rhodium dioxide, rhodium iv oxide, rho2
O=[Rh]=O
134.904
134.9
Rh 75.2 to 77.4%
Hygroscopic
250mg
12137-27-8
MFCD00016312
KZYDBKYFEURFNC-UHFFFAOYSA-N
dioxorhodium
82936
Powder
Odorless
Black
Chemical Identifiers
12137-27-8
134.904
KZYDBKYFEURFNC-UHFFFAOYSA-N
82936
O=[Rh]=O
O2Rh
MFCD00016312
rhodium iv oxide, rhodium oxide rho2, rhodium dioxide, rhodium iv oxide, rho2
dioxorhodium
Safety and Handling
Safety and Handling
TSCA : TSCA
SDS Documents