Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) sulfate, bright plating solution, Rh 5% (cont. Rh)

Chemical Identifiers

Chemical Name or Material Rhodium(III) sulfate
CAS 10489-46-0
Name Note Bright plating solution
Linear Formula Rh2(SO4)3
Formula Weight 493.99
MDL Number MFCD00016309
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12635NA

  SDS
alfa aesar™
12635NA
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12635NJ

  SDS
alfa aesar™
12635NJ
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 493.99
Chemical Name or Material Rhodium(III) sulfate
TSCA TSCA
Color Red-orange
Quantity 2g
Name Note Bright plating solution
Physical Form Liquid
Concentration Rh 5% (cont. Rh)
Linear Formula Rh2(SO4)3
Density 1.4g/mL
MDL Number MFCD00016309
CAS 10489-46-0
UN Number UN3264
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Acidic
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
RTECS Number FF9335000
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.