missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) sulfate, bright plating solution, Rh 5% (cont. Rh)

Manufacturer:  Alfa Aesar™ 12635NJ

 View more versions of this product

Catalog No. AA12635NJ



Specifications

Specifications

Rhodium(III) sulfate
10489-46-0
Rh2(SO4)3
UN3264
1.4g/mL
YWFDDXXMOPZFFM-UHFFFAOYSA-H
rhodium(3+);trisulfate
159290
Liquid
Red to Orange
Bright plating solution
O12Rh2S3
MFCD00016309
Rh 5% (cont. Rh)
rhodium iii sulfate solution, rhodium 3+ trisulfate, dirhodium trisulphate, rhodium sulfate, rhodium sulfate solution, acmc-1bntj, sulfuric acid, rhodium 3+ salt 3:2, rhodium iii sulfate, solution, dirhodium 3+ trisulfate, sulfuric acid,rhodium 3+ salt 3:2
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Rh+3].[Rh+3]
493.979
493.99
Acidic
0.5g

Chemical Identifiers

10489-46-0
493.979
YWFDDXXMOPZFFM-UHFFFAOYSA-H
159290
[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Rh+3].[Rh+3]
O12Rh2S3
MFCD00016309
rhodium iii sulfate solution, rhodium 3+ trisulfate, dirhodium trisulphate, rhodium sulfate, rhodium sulfate solution, acmc-1bntj, sulfuric acid, rhodium 3+ salt 3:2, rhodium iii sulfate, solution, dirhodium 3+ trisulfate, sulfuric acid,rhodium 3+ salt 3:2
rhodium(3+);trisulfate
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : FF9335000

TSCA : TSCA

Documents