Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) oxide pentahydrate, Premion™, 99.99% (metals basis), Rh 59% min

Chemical Identifiers

Chemical Name or Material Rhodium(III) oxide pentahydrate
CAS 39373-27-8
Purity Grade Premion™
Formula Weight 343.88 (253.81 Anhydrous)
MDL Number MFCD00149843
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1266701

  SDS
alfa aesar™
1266701
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1266703

  SDS
alfa aesar™
1266703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1266706

  SDS
alfa aesar™
1266706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 343.88 (253.81 Anhydrous)
Assay Percent Notes (metals basis)
Chemical Name or Material Rhodium(III) oxide pentahydrate
Purity Grade Premion™
MDL Number MFCD00149843
Assay 99.99%
TSCA TSCA
Quantity 0.25g
Physical Form Powder
Concentration Rh 59% min.
CAS 39373-27-8