missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) oxide pentahydrate, Premion™, 99.99% (metals basis), Rh 59% min

Manufacturer:  Alfa Aesar™ 1266706

 View more versions of this product

Catalog No. AA1266706



Specifications

Specifications

Rhodium(III) oxide pentahydrate
(metals basis)
MFCD00149843
XZIIPNYQAMUKBU-UHFFFAOYSA-N
oxo(oxorhodiooxy)rhodium;pentahydrate
45073257
Powder
Premion™
5g
39373-27-8
H10O8Rh2
rhodium iii oxide pentahydrate, o8rh2, rhodium-platinum oxide, rhodium iii oxide 5-hydrate, rhodium iii oxide pentahydrate, premion
O.O.O.O.O.O=[Rh]O[Rh]=O
343.883
343.88 (253.81 Anhydrous)
99.99%
Rh 59% min.

Chemical Identifiers

39373-27-8
343.883
XZIIPNYQAMUKBU-UHFFFAOYSA-N
45073257
O.O.O.O.O.O=[Rh]O[Rh]=O
H10O8Rh2
MFCD00149843
rhodium iii oxide pentahydrate, o8rh2, rhodium-platinum oxide, rhodium iii oxide 5-hydrate, rhodium iii oxide pentahydrate, premion
oxo(oxorhodiooxy)rhodium;pentahydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents