Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) oxide, anhydrous, 99.9% (metals basis), Rh 80.6% min

Chemical Identifiers

Chemical Name or Material Rhodium(III) oxide
CAS 12036-35-0
Name Note Anhydrous
Formula Weight 253.81
MDL Number MFCD00011205
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1181402

  SDS
alfa aesar™
1181402
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1181404

  SDS
alfa aesar™
1181404
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 253.81
Assay Percent Notes (metals basis)
Color Gray to black
Odor Odorless
Quantity 0.5g
Physical Form Powder
Concentration Rh 80.6% min.
Density 8.20g/mL
Sensitivity Hygroscopic
CAS 12036-35-0
Chemical Name or Material Rhodium(III) oxide
Melting Point 1100°C (dec.)
MDL Number MFCD00011205
TSCA TSCA
Assay 99.9%
Name Note Anhydrous