Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) nitrate solution, Rh 10-15% w/w (cont. Rh)

Chemical Identifiers

Chemical Name or Material Rhodium(III) nitrate solution
CAS 10139-58-9
Linear Formula Rh(NO3)3
Formula Weight 288.92
MDL Number MFCD00011202
UN Number UN3093
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12633NA

  SDS
alfa aesar™
12633NA
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12633NJ

  SDS
alfa aesar™
12633NJ
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 288.92
Chemical Name or Material Rhodium(III) nitrate solution
TSCA TSCA
Color Red-brown
Quantity 2g
Physical Form Liquid
Concentration Rh 10 to 15% w/w (cont. Rh)
Linear Formula Rh(NO3)3
MDL Number MFCD00011202
CAS 10139-58-9
UN Number UN3093
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H314-H318-H290-H302-H317
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Acidic
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
RTECS Number VI9316000
Safety and Handling

Danger

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H314-H318-H290-H302-H317
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.