missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Rhodium(III) nitrate solution, Rh 10-15% w/w (cont. Rh)

Manufacturer:  Alfa Aesar™ 12633NJ

 View more versions of this product

Catalog No. AA12633NJ



Specifications

Specifications

Rhodium(III) nitrate solution
N3O9Rh
MFCD00011202
Rh 10 to 15% w/w (cont. Rh)
VXNYVYJABGOSBX-UHFFFAOYSA-N
rhodium(3+);trinitrate
150190
Liquid
Red to Brown
10139-58-9
Rh(NO3)3
UN3093
rhodium nitrate, rhodium iii nitrate, rhodium trinitrate, nitric acid, rhodium 3+ salt, rhodium 3+ trinitrate, nitric acid, rhodium 3+ salt 3:1, nitric acid,rhodium 3+ salt 3:1, rhodiumtrinitrate, rhodium +3 nitrate, acmc-20ajsk
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Rh+3]
288.918
288.92
Acidic
0.5g

Chemical Identifiers

10139-58-9
288.918
VXNYVYJABGOSBX-UHFFFAOYSA-N
150190
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Rh+3]
N3O9Rh
MFCD00011202
rhodium nitrate, rhodium iii nitrate, rhodium trinitrate, nitric acid, rhodium 3+ salt, rhodium 3+ trinitrate, nitric acid, rhodium 3+ salt 3:1, nitric acid,rhodium 3+ salt 3:1, rhodiumtrinitrate, rhodium +3 nitrate, acmc-20ajsk
rhodium(3+);trinitrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318-H290-H302-H317
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.

RTECSNumber : VI9316000

TSCA : TSCA

Documents