Please login to your online account to display your discounted pricing

Rhodium(III)nitrate, solution, ca. 10% (w/v) Rh in 20-25 wt.% HNO3, ACROS Organics™

$377.37 - $377.37

Chemical Identifiers

Chemical Name or Material Rhodium(III)nitrate
Name Note Solution, ca.10% (w/w) Rh in 20-25 weight% HNO3
CAS 10139-58-9
CAS 7697-37-2
CAS 7732-18-5
Assay 20 to 25% HNO3 (w/w), 10 to 14% Rh (w/v)
Molecular Formula N3O9Rh
Linear Formula Rh(NO3)3
Formula Weight 288.92
MDL Number MFCD00011202
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC318980100

  SDS
thermo fisher scientific
AC318980100
Glass bottle 10mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC318980020

  SDS
thermo fisher scientific
318980020
Glass bottle 2mL Each for $377.37
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 288.92
Color Dark orange-brown to red-brown solution
Quantity 10mL
Assay 20 to 25% HNO3 (w/w), 10 to 14% Rh (w/v)
Linear Formula Rh(NO3)3
CAS 7732-18-5
Chemical Name or Material Rhodium(III)nitrate
Name Note Solution, ca.10% (w/w) Rh in 20-25 weight% HNO3
Packaging Glass bottle
Molecular Formula N3O9Rh
MDL Number MFCD00011202
CAS 10139-58-9
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
CAS 7697-37-2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.